8-Methyl-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

A163091-100MG A163091-250MG A163091-1G A163091-5G
103805-684EA 73 USD
103805-684 103843-942 76689-898 76689-900 77320-024 77320-026
8-Methyl-1,2,3,4-tetrahydroquinoline 95%
8-Methyl-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.2200 g/mol
Boiling Pt: 257.3 °C (760 mm Hg)
Flash Pt: 113.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00218100
CAS Number: 52601-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR