8-Methoxyquinoline-4-carbaldehyde 95%

Supplier: Ambeed
Warning

A396684-1G A396684-100MG A396684-250MG
103826-990EA 349 USD
103826-990 76841-456 77155-566 77155-568
8-Methoxyquinoline-4-carbaldehyde 95%
8-Methoxyquinoline-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06824369
CAS Number: 103854-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR