Monomethyl suberate 97%

Supplier: Ambeed
Warning

Synonyms: 8-Methoxy-8-oxooctanoic acid, Suberic Acid Monomethyl Ester, Methyl Hydrogen Suberate, Octanedioic Acid Monomethyl Ester

A119151-100G A119151-10G A119151-25G A119151-1G A119151-5G A119151-500G
103799-174EA 5 USD
103799-174 76781-618 76781-620 76781-622 77297-658 77297-660 77666-704
Monomethyl suberate 97%
Monomethyl suberate

Precaución: for Research Use Only

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 303.9 °C (760 mm Hg)
Melting Pt: 17…19 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004427
CAS Number: 3946-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR