8-Quinolinol 98%

Supplier: Ambeed
Danger

Synonyms: 8-Hydroxyquinoline

A257816-100G A257816-500G A257816-25G
103809-828EA 44 USD
103809-828 77000-876 77000-878 77000-880 77091-734
8-Quinolinol 98%
8-Quinolinol

Precaución: for Research Use Only

Formula: C₉H₇NO
Boiling Pt: 267 °C (760 mm Hg)
Melting Pt: 70…73 °C
Flash Pt: 267 °C
Storage Temperature: Ambient
MDL Number: MFCD00006807
CAS Number: 148-24-3
UN: 2811
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR