8-Fluoroquinoline 97%

Supplier: Ambeed
Warning

A125471-100G A125471-25G A125471-1G A125471-5G
103800-684EA 6 USD
103800-684 76971-850 76971-852 77298-052 77312-830
8-Fluoroquinoline 97%
8-Fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₆FN
MW: 147.1500 g/mol
Boiling Pt: 238.4 °C (760 mm Hg)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD01685514
CAS Number: 394-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR