8-Chloroquinolin-2-ol 97%

Supplier: Ambeed
Warning

A191752-10G A191752-1G A191752-25G A191752-5G
103816-396EA 5 USD
103816-396 103846-998 77029-830 77084-860 77084-862 77084-864
8-Chloroquinolin-2-ol 97%
8-Chloroquinolin-2-ol

Precaución: for Research Use Only

Formula: C₉H₆ClNO
Boiling Pt: 373.3±42.0 °C (760 mm Hg)
Melting Pt: 209…212 °C
Flash Pt: 179.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03427166
CAS Number: 23981-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR