8-Chloroimidazo[1,2-a]pyrazine 98%

Supplier: Ambeed
Warning

A466682-10G A466682-250MG A466682-1G A466682-5G A466682-100G
103827-982EA 13 USD
103827-982 103839-056 76695-408 76695-410 76842-968 76842-970 77626-934
8-Chloroimidazo[1,2-a]pyrazine 98%
8-Chloroimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09834910
CAS Number: 69214-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR