8-Cbz-2,4-dioxo-1,3,8-triazaspiro[4.5]decane 95%

Supplier: Ambeed
Warning

A934535-1G A934535-250MG
103841-416EA 294 USD
103841-416 76845-680 76993-948
8-Cbz-2,4-dioxo-1,3,8-triazaspiro[4.5]decane 95%
8-Cbz-2,4-dioxo-1,3,8-triazaspiro[4.5]decane

Precaución: for Research Use Only

Formula: C₁₅H₁₇N₃O₄
MW: 303.313 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18263009
CAS Number: 28121-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR