8-Bromoquinoline-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A232152-250MG A232152-5G A232152-1G
103810-816EA 34 USD
103810-816 103850-570 76815-182 77089-492 77303-932
8-Bromoquinoline-2-carboxylic acid 98%
8-Bromoquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MW: 252.06 g/mol
Flash Pt: 197.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690747
CAS Number: 914208-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR