8-Bromoquinazolin-4(1H)-one 95%

Supplier: Ambeed
Warning

A143019-100MG A143019-250MG A143019-1G A143019-5G A143019-10G
103803-232EA 48 USD
103803-232 76783-008 76783-010 77299-322 77316-478 77600-070
8-Bromoquinazolin-4(1H)-one 95%
8-Bromoquinazolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
Flash Pt: 185.1 °C
Storage Temperature: Ambient
MDL Number: MFCD10699396
CAS Number: 77150-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR