8-Bromocaprylic acid 98%

Supplier: Ambeed
Danger

A531421-25G A531421-5G A531421-10G A531421-1G
103828-570EA 7 USD
103828-570 103839-682 76843-696 76843-698 77197-336 77307-746
8-Bromocaprylic acid 98%
8-Bromocaprylic acid

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₆COOH
Boiling Pt: 316 °C (760 mm Hg)
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00004430
CAS Number: 17696-11-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR