8-Bromo-1-octanol 96%

Supplier: Ambeed
Warning

Synonyms: 8-Bromooctan-1-ol, 1-Bromo-8-hydroxyoctane

A157999-10G A157999-250MG A157999-1G A157999-25G A157999-5G A157999-100G A157999-500G
103805-082EA 6 USD
103805-082 103843-288 76973-396 76973-398 77300-310 77319-132 77319-134 77567-296 77611-516
8-Bromo-1-octanol 96%
8-Bromo-1-octanol

Precaución: For research use only.

Formula: BrH₂C(CH₂)₆CH₂OH
MW: 209.13 g/mol
Boiling Pt: 282.1 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010388
CAS Number: 50816-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR