8-Bromo-2-naphthol 95%

Supplier: Ambeed
Warning

Synonyms: 8-Bromonaphthalen-2-ol

A788870-100MG A788870-250MG A788870-1G A788870-5G
103841-006EA 399 USD
103841-006 76981-600 76981-602 77308-332 77308-334
8-Bromo-2-naphthol 95%
8-Bromo-2-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₇BrO
MW: 223.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656640
CAS Number: 7385-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR