8-Bromonaphthalen-1-ol 95%

Supplier: Ambeed
Warning

A127728-100MG A127728-250MG A127728-1G A127728-5G
103801-002EA 95 USD
103801-002 76870-606 76870-608 77313-264 77313-266
8-Bromonaphthalen-1-ol 95%
8-Bromonaphthalen-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₇BrO
MW: 223.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17012355
CAS Number: 62456-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR