8-Bromoisoquinoline 98%

Supplier: Ambeed
Warning

A120543-100G A120543-10G A120543-250MG A120543-25G A120543-1G A120543-5G
103799-956EA 9 USD
103799-956 77258-086 77258-088 77258-090 77258-092 77312-058 77312-060
8-Bromoisoquinoline 98%
8-Bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
Boiling Pt: 312.3 °C (760 mm Hg)
Melting Pt: 80.5 °C
Flash Pt: 111.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04973298
CAS Number: 63927-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR