8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine 98%

Supplier: Ambeed
Warning

Synonyms: 8-Bromo-7-(but-2-yn-1-yl)-3-methyl-1H-purine-2,6(3H,7H)-dione, 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione

A479467-1G A479467-5G A479467-10G A479467-250MG
103828-112EA 9 USD
103828-112 103839-186 76843-122 76843-124 76877-816 76877-818
8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine 98%
8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₄O₂
MW: 297.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22126091
CAS Number: 666816-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR