8-Bromo-6-chloroimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A113423-100MG A113423-250MG A113423-1G
103798-230EA 15 USD
103798-230 103824-072 76982-848 76982-850 77310-886
8-Bromo-6-chloroimidazo[1,2-a]pyridine 97%
8-Bromo-6-chloroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD12828062
CAS Number: 957187-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR