8-Bromo-5-nitroquinoline 98%

Supplier: Ambeed
Warning

A235297-100MG A235297-10G A235297-250MG A235297-25G A235297-5G A235297-1G
103811-000EA 23 USD
103811-000 103850-764 77031-172 77031-174 77031-176 77089-770 77089-772 77304-006
8-Bromo-5-nitroquinoline 98%
8-Bromo-5-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrN₂O₂
MW: 253.0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00509069
CAS Number: 139366-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR