8-Bromo-5-fluoroquinoline 95%

Supplier: Ambeed
Warning

A116474-100MG A116474-250MG A116474-1G A116474-5G
103798-738EA 46 USD
103798-738 103824-604 77026-138 77026-140 77311-410 77311-412
8-Bromo-5-fluoroquinoline 95%
8-Bromo-5-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MW: 226.0451 g/mol
Boiling Pt: 294.2±20.0 °C (760 mm Hg)
Flash Pt: 131.7 °C
Storage Temperature: Ambient
MDL Number: MFCD09907839
CAS Number: 917251-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR