8-Bromo-4-chloroquinazoline 97%
Supplier: Ambeed
|
Warning
|
A207740-100MG
A207740-10G
A207740-250MG
A207740-1G
103817-816EA
78
USD
103817-816
103848-500
76785-580
76785-582
76785-584
77086-786
8-Bromo-4-chloroquinazoline 97%
8-Bromo-4-chloroquinazoline
Precaución: for Research Use Only
Formula:
C₈H₄BrClN₂ MW: 243.4900 g/mol Boiling Pt: 339.7 °C (760 mm Hg) Flash Pt: 159.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09738589 CAS Number: 125096-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...