8-Bromo-2-methylquinoline 97%
Supplier: Ambeed
|
Warning
|
A383087-1G
A383087-25G
A383087-5G
A383087-100MG
A383087-250MG
103826-730EA
13
USD
103826-730
103837-766
76841-088
76841-090
76841-092
76978-842
76978-844
8-Bromo-2-methylquinoline 97%
8-Bromo-2-methylquinoline
Precaución: For research use only
Formula:
C₁₀H₈BrN MW: 222.0812 g/mol Boiling Pt: 299.7 °C (760 mm Hg) Melting Pt: 63…65 °C Flash Pt: 135 °C Storage Temperature: Ambient |
MDL Number:
MFCD04966997 CAS Number: 61047-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...