8-Bromo-2-methoxy-1,5-naphthyridine 97%

Supplier: Ambeed
Warning

A148276-1G A148276-5G
103803-906EA 62 USD
103803-906 103842-022 77317-464 77317-466
8-Bromo-2-methoxy-1,5-naphthyridine 97%
8-Bromo-2-methoxy-1,5-naphthyridine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09027616
CAS Number: 881658-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR