8-Bromo-2-chloroquinoline 98%

Supplier: Ambeed
Warning

A321453-10G A321453-250MG A321453-1G A321453-25G A321453-5G
103825-274EA 11 USD
103825-274 103836-208 76788-064 76788-066 76839-106 76839-108 76839-110
8-Bromo-2-chloroquinoline 98%
8-Bromo-2-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
Storage Temperature: Refrigerator
MDL Number: MFCD08741410
CAS Number: 163485-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR