8-Bromo-1-naphthoic acid 95%
Supplier: Ambeed
|
Warning
|
A269916-10G
A269916-250MG
A269916-1G
A269916-5G
103812-982EA
18
USD
103812-982
103852-888
76801-954
76801-956
77305-040
77305-042
8-Bromo-1-naphthoic acid 95%
8-Bromo-1-naphthoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₇BrO₂ MW: 251.0761 g/mol Boiling Pt: 421.2 °C (760 mm Hg) Melting Pt: 169…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00029357 CAS Number: 1729-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...