8-Bromo-1,6-naphthyridin-5-amine 97%

Supplier: Ambeed
Warning

A159951-10G A159951-1G A159951-100MG
103805-304EA 167 USD
103805-304 103843-532 77191-042 77319-438 77746-093
8-Bromo-1,6-naphthyridin-5-amine 97%
8-Bromo-1,6-naphthyridin-5-amine

Precaución: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₈H₆BrN₃
Boiling Pt: 360.1±37.0 °C (760 mm Hg)
Flash Pt: 171.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22396116
CAS Number: 1820686-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR