8-Bromo-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: Ambeed
Warning

A276454-100MG A276454-250MG A276454-1G A276454-5G
103813-304EA 46 USD
103813-304 103853-238 76816-312 76816-314 77093-094 77093-096
8-Bromo-1,2,3,4-tetrahydroisoquinoline 97%
8-Bromo-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀BrN
MW: 212.09 g/mol
Boiling Pt: 294.3 °C (760 mm Hg)
Flash Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10001500
CAS Number: 75416-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR