8-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 8-Bromo-[1,2,4]triazolo[1,5-a]pyridine

A271779-250MG A271779-5G
103813-064EA 32 USD
103813-064 103852-976 76692-850 77092-760
8-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
8-Bromo[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.03 g/mol
Melting Pt: 152…157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10699199
CAS Number: 868362-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR