8-Boc-2,8-Diazaspiro[4.5]decane 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2,8-diazaspiro[4.5]decane-8-carboxylate, 2,8-Diazaspiro[4.5]decane-8-carboxylic acid-tert-butyl ester

A230165-1G A230165-100MG A230165-10G A230165-250MG A230165-25G
103819-528EA 189 USD
103819-528 103850-428 77089-288 77249-548 77249-550 77249-552 77746-344
8-Boc-2,8-Diazaspiro[4.5]decane 98%
8-Boc-2,8-Diazaspiro[4.5]decane

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MW: 240.342 g/mol
Boiling Pt: 337 °C (760 mm Hg)
Flash Pt: 157.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09608078
CAS Number: 236406-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR