N-Benzyltropinone 98%

Supplier: Ambeed
Warning

Synonyms: 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one, (1R,5S)-8-Benzyl-8-azabicyclo[3.2.1]octan-3-one

A126257-100G A126257-10G A126257-1G A126257-25G A126257-5G
103800-782EA 6 USD
103800-782 76782-016 76782-018 77312-980 77312-982 77312-984
N-Benzyltropinone 98%
N-Benzyltropinone

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO
MW: 215.30 g/mol
Boiling Pt: 342.9 °C (760 mm Hg)
Melting Pt: 28…29 °C
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00151640
CAS Number: 28957-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR