3,3-Tetramethyleneglutarimide 97%

Supplier: Ambeed
Danger

Synonyms: 8-Azaspiro[4.5]decane-7,9-dione, Tetramethylene glutarimide

A319198-25G A319198-1G
103825-192EA 7 USD
103825-192 103836-124 76838-974 77306-196
3,3-Tetramethyleneglutarimide 97%
3,3-Tetramethyleneglutarimide

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Boiling Pt: 350.7 °C (760 mm Hg)
Melting Pt: 153…155 °C
Flash Pt: 162.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00023871
CAS Number: 1075-89-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR