8-Aminoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: Quinolin-8-ylamine

A104160-100G A104160-10G A104160-25G A104160-1G A104160-5G
103796-662EA 9 USD
103796-662 103822-376 77270-374 77270-376 77270-378 77309-238 77309-240
8-Aminoquinoline 97%
8-Aminoquinoline

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 289.8 °C (760 mm Hg)
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00006809
CAS Number: 578-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR