8-Aminocaprylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
ω-Aminooctoic acid, 8-Aminooctanoic acid, ω-Aminocaprylic acid, ω-Aminooctanoic acid, 8-Aminooctoic acid
A114797-10G
A114797-250MG
A114797-25G
A114797-5G
A114797-1G
103798-478EA
6
USD
103798-478
103824-320
77189-284
77189-286
77189-288
77297-326
77311-148
8-Aminocaprylic acid 97%
8-Aminocaprylic acid
Precaución: for Research Use Only
Formula:
C₈H₁₇NO₂ MW: 159.23 g/mol Melting Pt: 194 °C Flash Pt: 126.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008245 CAS Number: 1002-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...