6-Methoxyquinolin-8-amine 98%

Supplier: Ambeed
Warning

Synonyms: 8-Amino-6-methoxyquinoline

A168793-100G A168793-10G A168793-250MG A168793-1G A168793-5G
103806-318EA 15 USD
103806-318 103844-572 76690-116 76690-118 76690-120 77081-638 77300-944
6-Methoxyquinolin-8-amine 98%
6-Methoxyquinolin-8-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Boiling Pt: 361.8±27.0 °C (760 mm Hg)
Melting Pt: 41 °C
Flash Pt: 172.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672902
CAS Number: 90-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR