8-(Trifluoromethyl)quinazolin-2-amine 97%

Supplier: Ambeed
Warning

A794423-1G A794423-100MG
103841-034EA 1568 USD
103841-034 76845-278 77268-198
8-(Trifluoromethyl)quinazolin-2-amine 97%
8-(Trifluoromethyl)quinazolin-2-amine

Precaución: for Research Use Only

Formula: C₉H₆F₃N₃
MW: 213.16 g/mol
Boiling Pt: 359.4±52.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD18157724
CAS Number: 1258652-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR