8-(tert-Butoxy)-8-oxooctanoic acid 97%

Supplier: Ambeed
Warning

A257544-10G A257544-250MG A257544-1G A257544-5G
103812-318EA 47 USD
103812-318 103852-186 77098-760 77098-762 77304-710 77304-712
8-(tert-Butoxy)-8-oxooctanoic acid 97%
8-(tert-Butoxy)-8-oxooctanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₄
MW: 230.3 g/mol
Boiling Pt: 324.8 °C (760 mm Hg)
Flash Pt: 113.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD28505597
CAS Number: 234081-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR