8-(Bromomethyl)quinoline 95%

Supplier: Ambeed
Danger

A179849-1G A179849-5G A179849-100MG A179849-10G A179849-250MG
103807-480EA 24 USD
103807-480 103845-834 77083-306 77083-308 77163-840 77163-842 77163-844
8-(Bromomethyl)quinoline 95%
8-(Bromomethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.0800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00047621
CAS Number: 7496-46-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR