3,4-Epoxycyclohexylmethyl-3,4-epoxycyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 7-Oxabicyclo[4.1.0]heptan-3-ylmethyl 7-oxabicyclo[4.1.0]heptane-3-carboxylate

A131652-100G A131652-10G A131652-500G A131652-1000G A131652-25G
103808-432EA 92 USD
103808-432 76688-648 76688-650 76688-652 77314-122 77314-124
3,4-Epoxycyclohexylmethyl-3,4-epoxycyclohexanecarboxylate 97%
3,4-Epoxycyclohexylmethyl-3,4-epoxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₄
MW: 252.31 g/mol
Boiling Pt: 363.4 °C (760 mm Hg)
Melting Pt: …37 °C
Flash Pt: 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00440900
CAS Number: 2386-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR