7-Oxabicyclo[4.1.0]heptan-2-one 95%

Supplier: Ambeed
Warning

A249498-10G A249498-250MG A249498-1G A249498-5G
103811-876EA 32 USD
103811-876 103851-714 76815-624 76815-626 77304-486 77304-488
7-Oxabicyclo[4.1.0]heptan-2-one 95%
7-Oxabicyclo[4.1.0]heptan-2-one

Precaución: for Research Use Only

Formula: C₆H₈O₂
Boiling Pt: 204.5 °C (760 mm Hg)
Flash Pt: 96.11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192371
CAS Number: 6705-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR