7-Oxa-2-azaspiro[3.5]nonane hydrochloride 95%

Supplier: Ambeed
Warning

A256260-100MG A256260-250MG
103812-256EA 109 USD
103812-256 103852-120 76988-044 76988-046
7-Oxa-2-azaspiro[3.5]nonane hydrochloride 95%
7-Oxa-2-azaspiro[3.5]nonane hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO·ClH
MW: 163.645 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22421803
CAS Number: 1417633-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR