7-Octyn-1-ol 96%
Supplier: Ambeed
|
Warning
|
A494927-100G
A494927-10G
A494927-25G
A494927-5G
A494927-1G
103828-246EA
9
USD
103828-246
103839-326
76695-664
76695-666
76843-292
76843-294
77307-594
7-Octyn-1-ol 96%
7-Octyn-1-ol
Precaución: for Research Use Only
Formula:
C₈H₁₄O Boiling Pt: 191.5 °C (760 mm Hg) Flash Pt: 116.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632137 CAS Number: 871-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...