7-Nitroquinoline 98%

Supplier: Ambeed
Warning

A722991-100G A722991-10G A722991-250MG A722991-1G A722991-25G
103840-756EA 22 USD
103840-756 76697-526 76697-528 76697-530 76844-976 76844-978
7-Nitroquinoline 98%
7-Nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
MW: 174.1561 g/mol
Melting Pt: 129…130 °C
Flash Pt: 156.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00234498
CAS Number: 613-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR