7-Nitro-2(1H)-quinoxalinone 97%

Supplier: Ambeed
Warning

A309018-100MG A309018-250MG A309018-1G A309018-5G
103814-584EA 88 USD
103814-584 103835-762 76787-892 76787-894 77306-004 77306-006
7-Nitro-2(1H)-quinoxalinone 97%
7-Nitro-2(1H)-quinoxalinone

Precaución: for Research Use Only

Formula: C₈H₅N₃O₃
Melting Pt: 275 °C
Storage Temperature: Ambient
MDL Number: MFCD09999191
CAS Number: 89898-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR