7-Nitroindole 98%

Supplier: Ambeed
Warning

A260818-100G A260818-10G A260818-1G A260818-25G A260818-5G A260818-500G
103812-508EA 5 USD
103812-508 103852-394 76692-702 76692-704 77091-986 77091-988 77304-806 77626-362
7-Nitroindole 98%
7-Nitroindole

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Boiling Pt: 362.6 °C (760 mm Hg)
Melting Pt: 94…98 °C
Flash Pt: 173.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00005683
CAS Number: 6960-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR