7-Nitro-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydro-7-nitroisoquinoline

A315838-5G A315838-10G A315838-1G A315838-250MG
103825-086EA 19 USD
103825-086 103836-008 76838-838 77167-214 77306-138 77683-938
7-Nitro-1,2,3,4-tetrahydroisoquinoline 97%
7-Nitro-1,2,3,4-tetrahydroisoquinoline

Precaución: For research use only.

Formula: C₉H₁₀N₂O₂
MW: 178.1879 g/mol
Boiling Pt: 319.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04973400
CAS Number: 42923-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR