7-Methylisatin 97%

Supplier: Ambeed
Warning

Synonyms: 7-Methylindoline-2,3-dione

A125670-10G A125670-250MG A125670-25G A125670-1G A125670-5G
103800-710EA 6 USD
103800-710 76688-380 76688-382 77298-082 77312-856 77312-858
7-Methylisatin 97%
7-Methylisatin

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.1574 g/mol
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022797
CAS Number: 1127-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR