7-Methylindole-3-carboxyaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 7-Methyl-1H-indole-3-carbaldehyde

A131482-100MG A131482-10G A131482-250MG A131482-1G A131482-25G A131482-5G
103801-588EA 23 USD
103801-588 76972-090 76972-092 76972-094 77298-488 77298-490 77298-492
7-Methylindole-3-carboxyaldehyde 98%
7-Methylindole-3-carboxyaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO
Boiling Pt: 341 °C (760 mm Hg)
Melting Pt: 206…208 °C
Storage Temperature: Freezer
MDL Number: MFCD00047170
CAS Number: 4771-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR