7-Methyl-6-nitro-1H-indazole 95%

Supplier: Ambeed
Warning

A583472-1G A583472-5G
103828-876EA 18 USD
103828-876 76844-156 77886-418
7-Methyl-6-nitro-1H-indazole 95%
7-Methyl-6-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
Boiling Pt: 379.7±22.0 °C (760 mm Hg)
Melting Pt: 224…226 °C
Flash Pt: 183.4 °C
Storage Temperature: Ambient
MDL Number: MFCD16620021
CAS Number: 208457-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR