7-Methyl-1H-indole-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A107177-100MG
A107177-250MG
103797-146EA
42
USD
103797-146
103822-908
76796-606
76796-608
7-Methyl-1H-indole-2-carboxylic acid 97%
7-Methyl-1H-indole-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Boiling Pt: 413.3 °C (760 mm Hg) Melting Pt: 169…175 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00047168 CAS Number: 18474-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...