7-Methylindole 98%

Supplier: Ambeed
Warning

A134573-100G A134573-10G A134573-1G A134573-25G A134573-5G
103802-022EA 5 USD
103802-022 76688-758 76688-760 77314-782 77314-784 77314-786
7-Methylindole 98%
7-Methylindole

Precaución: for Research Use Only

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 266 °C (760 mm Hg)
Melting Pt: 80…84 °C
Flash Pt: 266 °C
Storage Temperature: Ambient
MDL Number: MFCD00005684
CAS Number: 933-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR