7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene 95%

Supplier: Ambeed
Danger

Synonyms: MTBD, 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine

A115435-100MG A115435-10G A115435-250MG A115435-25G A115435-1G A115435-5G
103798-564EA 24 USD
103798-564 103824-420 77189-344 77189-346 77189-348 77189-350 77311-238 77311-240
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene 95%
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene

Precaución: for Research Use Only

Formula: C₈H₁₅N₃
MW: 153.23 g/mol
Boiling Pt: 298.7 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043004
CAS Number: 84030-20-6
UN: 3267
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR